missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZSCAN4 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-05600-20ul
This item is not returnable.
View return policy
Description
ZSCAN4 Polyclonal antibody specifically detects ZSCAN4 in Mouse samples. It is validated for Western Blot
Specifications
| ZSCAN4 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| FLJ35105, MGC126787, zinc finger and SCAN domain containing 4, Zinc finger protein 494MGC126789, ZNF494zinc finger and SCAN domain-containing protein 4 | |
| A synthetic peptide corresponding to a sequence within amino acids 400-506 of mouse ZSCAN4 (NP_001013787.1). CSRMFKHARSLSSHQRTHLNKKSELLCVTCQKMFKRVSDRRTHEIIHMPEKPFKCSTCEKSFSHKTNLKSHEMIHTGEMPYVCSLCSRRFRQSSTYHRHLRNYHRSD | |
| 20 μg | |
| Primary | |
| Mouse | |
| Purified |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 201516 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction