missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZSCAN4 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 188.00 - € 463.00
Specifications
| Antigen | ZSCAN4 |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18609346
|
Novus Biologicals
NBP3-05600-100ul |
100 μg |
€ 463.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18695547
|
Novus Biologicals
NBP3-05600-20ul |
20 μg |
€ 188.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ZSCAN4 Polyclonal antibody specifically detects ZSCAN4 in Mouse samples. It is validated for Western BlotSpecifications
| ZSCAN4 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Mouse | |
| FLJ35105, MGC126787, zinc finger and SCAN domain containing 4, Zinc finger protein 494MGC126789, ZNF494zinc finger and SCAN domain-containing protein 4 | |
| A synthetic peptide corresponding to a sequence within amino acids 400-506 of mouse ZSCAN4 (NP_001013787.1). CSRMFKHARSLSSHQRTHLNKKSELLCVTCQKMFKRVSDRRTHEIIHMPEKPFKCSTCEKSFSHKTNLKSHEMIHTGEMPYVCSLCSRRFRQSSTYHRHLRNYHRSD | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS with 50% glycerol, pH7.3. | |
| 201516 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title