accessibility menu, dialog, popup

UserName

CAS RN 120928-09-8

H3CCH3H3CONN

CAS RN 120928-09-8

IUPAC Name: 4-[2-(4-tert-butylphenyl)ethoxy]quinazoline
Synonyms: fenazaquin
Molecular Weight (g/mol): 306.41
Molecular Formula: C20H22N2O
InChi Key: DMYHGDXADUDKCQ-UHFFFAOYSA-N
SMILES: CC(C)(C)C1=CC=C(CCOC2=C3C=CC=CC3=NC=N2)C=C1
Molecular Weight (g/mol) 
  • (3)
  • (1)
  • (3)
Concentration 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Solvent or Matrix 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (3)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)