CAS RN 1234356-69-4
CAS RN 1234356-69-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Derazantinib, MedChemExpress
MedChemExpress Derazantinib (ARQ-087) is an orally bioavailable, ATP competitive tyrosine kinase inhibitor; exhibits potent activity against FGFR1-3 chondrocytes with IC50s of 4.5, 1.8, and 4.5 nM, respectively.