accessibility menu, dialog, popup

UserName

CAS RN 132274-70-5

NNO

CAS RN 132274-70-5

IUPAC Name: 1-phenyl-1H-pyrazole-5-carbaldehyde
Synonyms: 2-phenylpyrazole-3-carbaldehyde
Molecular Weight (g/mol): 172.19
Molecular Formula: C10H8N2O
InChi Key: OQORFMABOZEDBL-UHFFFAOYSA-N
SMILES: O=CC1=CC=NN1C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results