CAS RN 13472-59-8
CAS RN 13472-59-8
IUPAC Name:
3-bromo-2-methoxypyridine
Synonyms:
3-bromo-2-methoxypyridine
Molecular Weight (g/mol):
188.02
Molecular Formula:
C6H6BrNO
InChi Key:
PORGLLGXCAQORO-UHFFFAOYSA-N
SMILES:
COC1=C(Br)C=CC=N1
1
–
2
of
2
results
2
3-Bromo-2-methoxypyridine, 95%, Thermo Scientific™
CAS: 13472-59-8 Molecular Formula: C6H6BrNO Molecular Weight (g/mol): 188.02 MDL Number: MFCD03095349 InChI Key: PORGLLGXCAQORO-UHFFFAOYSA-N Synonym: 2-methoxy-3-bromopyridine,3-bromo-2-methoxy-pyridine,3-brown-2-methoxypyridine,pyridine, 3-bromo-2-methoxy,tspp decahydrate,pubchem1180,2-methoxybromopyridine,acmc-209bwq,ksc494g7f,3-bromanyl-2-methoxy-pyridine PubChem CID: 12533390 IUPAC Name: 3-bromo-2-methoxypyridine SMILES: COC1=C(Br)C=CC=N1