missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-2-methoxypyridine, 95%, Thermo Scientific™

Product Code. 11312329
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11312329 5 g 5g
11302329 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11312329 Supplier Thermo Scientific Alfa Aesar Supplier No. H27279.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is employed in the facile synthesis of 2-unsubstituted benzofuran-3-carboxylates using diazo(trimethylsilyl)methyl magnesium bromide.

Solubility
Not miscible or difficult to mix in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13472-59-8
Molecular Formula C6H6BrNO
Molecular Weight (g/mol) 188.02
MDL Number MFCD03095349
InChI Key PORGLLGXCAQORO-UHFFFAOYSA-N
Synonym 2-methoxy-3-bromopyridine, 3-bromo-2-methoxy-pyridine, 3-brown-2-methoxypyridine, pyridine, 3-bromo-2-methoxy, tspp decahydrate, pubchem1180, 2-methoxybromopyridine, acmc-209bwq, ksc494g7f, 3-bromanyl-2-methoxy-pyridine
PubChem CID 12533390
IUPAC Name 3-bromo-2-methoxypyridine
SMILES COC1=C(Br)C=CC=N1

Specifications

Density 1.59
Flash Point 93°C (199°F)
Refractive Index 1.5635
Quantity 5 g
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 188.02
Percent Purity 95%
Chemical Name or Material 3-Bromo-2-methoxypyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.