accessibility menu, dialog, popup

UserName

CH3OOH3C(E)

CAS RN 13894-63-8

IUPAC Name: methyl (2E)-hex-2-enoate
Synonyms: methyl hexenoate
Molecular Weight (g/mol): 128.17
Molecular Formula: C7H12O2
InChi Key: GFUGBRNILVVWIE-AATRIKPKSA-N
SMILES: CCC\C=C\C(=O)OC
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results