CAS RN 1394969-56-2
CAS RN 1394969-56-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
[1,2,4]Triazolo[1,5-a]pyridin-6-ol, TRC
CAS: 1394969-56-2 Molecular Formula: C6H5N3O Molecular Weight (g/mol): 135.12 IUPAC Name: [1,2,4]triazolo[1,5-a]pyridin-6-ol SMILES: Oc1ccc2ncnn2c1