CAS RN 1446486-33-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
3
of
3
results
1
(E)-9-(Prop-1-enyl)-9H-purin-6-amine, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2
9-Propenyladenine, TRC
CAS: 1446486-33-4 Molecular Formula: C8 H9 N5 Molecular Weight (g/mol): 175.19 Synonym: 9-(1E)-1-Propen-1-yl-9H-purin-6-amine IUPAC Name: 9-[(E)-prop-1-enyl]purin-6-amine SMILES: C\C=C\n1cnc2c(N)ncnc12
3
9-Propenyladenine, MedChemExpress
MedChemExpress 9-Propenyladenine is a mutagenic impurity in tenofovir disoproxil fumarate. Tenofovir is an antiretroviral drug known as nucleotide analogue reverse transcriptase inhibitors, which block reverse transcriptase, a crucial virus enzyme in HIV-1 and HBV.