accessibility menu, dialog, popup

UserName

CAS RN 1658-56-6

NaOSOONHNO(E)

CAS RN 1658-56-6

IUPAC Name: sodium 4-{2-[(1E)-2-oxo-1,2-dihydronaphthalen-1-ylidene]hydrazin-1-yl}naphthalene-1-sulfonate
Synonyms: acid red 88
Molecular Weight (g/mol): 400.38
Molecular Formula: C20H13N2NaO4S
InChi Key: YCRBPWIFISBYGJ-QPNALZDCSA-M
SMILES: [Na+].[O-]S(=O)(=O)C1=C2C=CC=CC2=C(N\N=C2\C(=O)C=CC3=CC=CC=C23)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results