CAS RN 18640-58-9
CAS RN 18640-58-9
IUPAC Name:
1-(4-bromo-3-nitrophenyl)ethan-1-one
Synonyms:
1-(4-bromo-3-nitrophenyl)ethanone
Molecular Weight (g/mol):
244.04
Molecular Formula:
C8H6BrNO3
InChi Key:
YFVOFFKNHQTQQE-UHFFFAOYSA-N
SMILES:
CC(=O)C1=CC=C(Br)C(=C1)[N+]([O-])=O
1
–
2
of
2
results
1
4'-Bromo-3'-nitroacetophenone, 99%
CAS: 18640-58-9 Molecular Formula: C8H6BrNO3 Molecular Weight (g/mol): 244.04 MDL Number: MFCD00016985 InChI Key: YFVOFFKNHQTQQE-UHFFFAOYSA-N Synonym: 1-4-bromo-3-nitrophenyl ethanone,4'-bromo-3'-nitroacetophenone,1-4-bromo-3-nitrophenyl ethan-1-one,4-bromo-3-nitroacetophenone,4-bromo-3'-nitroacetophenone,ethanone, 1-4-bromo-3-nitrophenyl,acmc-209eny,3-nitro-4-bromoacetophenone,4-bromo-3-nitro acetophenone,3'-nitro-4'-bromoacetophenone PubChem CID: 87737 IUPAC Name: 1-(4-bromo-3-nitrophenyl)ethanone SMILES: CC(=O)C1=CC=C(Br)C(=C1)[N+]([O-])=O