accessibility menu, dialog, popup

UserName

CAS RN 19781-81-8

CH3H3COH

CAS RN 19781-81-8

IUPAC Name: hept-4-yn-2-ol
Synonyms: hept-4-yn-2-ol
Molecular Weight (g/mol): 112.17
Molecular Formula: C7H12O
InChi Key: CXVIEBMEWKSONY-UHFFFAOYNA-N
SMILES: CCC#CCC(C)O
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)