accessibility menu, dialog, popup

UserName

CAS RN 198141-12-7

IUPAC Name: 5H,6H,7H,8H,9H-pyrido[3,2-b]indol-9-one
Synonyms: 5H,6H,7H,8H-pyrido[3,2-b]indol-9-one
Molecular Weight (g/mol): 186.21
Molecular Formula: C11H10N2O
InChi Key: FATNUJIDKOPJRF-UHFFFAOYSA-N
SMILES: O=C1CCCC2=C1C1=NC=CC=C1N2
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results