accessibility menu, dialog, popup

UserName

CAS RN 198470-85-8

NaH3CONSOOCH3ON

CAS RN 198470-85-8

IUPAC Name: sodium [4-(5-methyl-3-phenyl-1,2-oxazol-4-yl)benzenesulfonyl](propanoyl)azanide
Synonyms: sodium [4-(5-methyl-3-phenyl-1,2-oxazol-4-yl)benzenesulfonyl](propanoyl)azanide
Molecular Weight (g/mol): 392.4
Molecular Formula: C19H17N2NaO4S
InChi Key: HQPVVKXJNZEAFW-UHFFFAOYSA-M
SMILES: [Na+].CCC(=O)[N-]S(=O)(=O)C1=CC=C(C=C1)C1=C(C)ON=C1C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (5)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results