CAS RN 20348-09-8
CAS RN 20348-09-8
IUPAC Name:
2H,3H,4H-pyrido[3,2-b][1,4]oxazin-3-one
Synonyms:
2H,4H-pyrido[3,2-b][1,4]oxazin-3-one
Molecular Weight (g/mol):
150.14
Molecular Formula:
C7H6N2O2
InChi Key:
ANHQLUBMNSSPBV-UHFFFAOYSA-N
SMILES:
O=C1COC2=CC=CN=C2N1
1
–
1
of
1
results
1
2H-Pyrido[3,2-b]-1,4-oxazin-3(4H)-one, 98%, Thermo Scientific™
CAS: 20348-09-8 Molecular Formula: C7H6N2O2 Molecular Weight (g/mol): 150.137 MDL Number: MFCD00006697 InChI Key: ANHQLUBMNSSPBV-UHFFFAOYSA-N Synonym: 2h-pyrido 3,2-b 1,4 oxazin-3 4h-one,2h-pyrido 3,2-b-1,4-oxazin-3 4h-one,4h-pyrido 3,2-b 1,4 oxazin-3-one,2h,3h,4h-pyrido 3,2-b 1,4 oxazin-3-one,2h,4h-pyrido 3,2-b 1,4 oxazin-3-one,2h,4h-pyridino 2,3-e 1,4-oxazin-3-one,pyridoboxazinone,acmc-20ahw7,2h-pyrido 3,2-b 1,4 oxazin-3-one PubChem CID: 88499 IUPAC Name: 4H-pyrido[3,2-b][1,4]oxazin-3-one SMILES: C1C(=O)NC2=C(O1)C=CC=N2