Learn More
2H-Pyrido[3,2-b]-1,4-oxazin-3(4H)-one, 98%, Thermo Scientific™
Product Code. 11358185
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11358185 5 g 5g
11348185 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11358185 Supplier Thermo Scientific Alfa Aesar Supplier No. L08473.06
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
missing translation for 'validMainframeMsg'
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 20348-09-8
Molecular Formula C7H6N2O2
Molecular Weight (g/mol) 150.137
MDL Number MFCD00006697
InChI Key ANHQLUBMNSSPBV-UHFFFAOYSA-N
Synonym 2h-pyrido 3,2-b 1,4 oxazin-3 4h-one, 2h-pyrido 3,2-b-1,4-oxazin-3 4h-one, 4h-pyrido 3,2-b 1,4 oxazin-3-one, 2h,3h,4h-pyrido 3,2-b 1,4 oxazin-3-one, 2h,4h-pyrido 3,2-b 1,4 oxazin-3-one, 2h,4h-pyridino 2,3-e 1,4-oxazin-3-one, pyridoboxazinone, acmc-20ahw7, 2h-pyrido 3,2-b 1,4 oxazin-3-one
PubChem CID 88499
IUPAC Name 4H-pyrido[3,2-b][1,4]oxazin-3-one
SMILES C1C(=O)NC2=C(O1)C=CC=N2
Specifications
Melting Point 204°C to 208°C
Quantity 5 g
Beilstein 1074010
Formula Weight 150.14
Percent Purity 98%
Chemical Name or Material 2H-Pyrido[3,2-b]-1,4-oxazin-3(4H)-one