CAS RN 21172-43-0
CAS RN 21172-43-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Methanesulfonic Acid 3-(3-Trifluoromethylphenyl)propyl Ester, TRC
CAS: 21172-43-0 Molecular Formula: C11 H13 F3 O3 S Molecular Weight (g/mol): 282.28 Synonym: Benzenepropanol, 3-(trifluoromethyl)-, 1-methanesulfonate, 1-Propanol, 3-(α,α,α-trifluoro-m-tolyl)-, methanesulfonate (8CI),Benzenepropanol, 3-(trifluoromethyl)-, methanesulfonate (9CI),3-(3-Trifluoromethylphenyl)propyl methanesulfonate,Methanesulfonic acid 3-(3-trifluoromethylphenyl)propyl ester,3-(trifluoromethyl)Benzenepropanal, Methanesulfonate,1-Propanol, 3-(α,α,α-trifluoro-m-tolyl)-, Methanesulfonate,Benzenepropanol, 3-(trifluoromethyl)-, 1-Methanesulfonate IUPAC Name: 3-[3-(trifluoromethyl)phenyl]propyl methanesulfonate SMILES: CS(=O)(=O)OCCCc1cccc(c1)C(F)(F)F