accessibility menu, dialog, popup

UserName

NH2

CAS RN 2243-47-2

IUPAC Name: [1,1'-biphenyl]-3-amine
Synonyms: [1,1'-biphenyl]-3-amine
Molecular Weight (g/mol): 169.23
Molecular Formula: C12H11N
InChi Key: MUNOBADFTHUUFG-UHFFFAOYSA-N
SMILES: NC1=CC=CC(=C1)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results