CAS RN 252004-45-8
CAS RN 252004-45-8
IUPAC Name:
4-chloro-2,6-difluorobenzaldehyde
Synonyms:
4-chloro-2,6-difluorobenzaldehyde
Molecular Weight (g/mol):
176.55
Molecular Formula:
C7H3ClF2O
InChi Key:
GTTVSBCPMJQRSP-UHFFFAOYSA-N
SMILES:
FC1=CC(Cl)=CC(F)=C1C=O
1
–
2
of
2
results
1
4-Chloro-2,6-difluorobenzaldehyde, 97%, Thermo Scientific™
CAS: 252004-45-8 Molecular Formula: C7H3ClF2O Molecular Weight (g/mol): 176.547 MDL Number: MFCD03094460 InChI Key: GTTVSBCPMJQRSP-UHFFFAOYSA-N PubChem CID: 2773512 IUPAC Name: 4-chloro-2,6-difluorobenzaldehyde SMILES: C1=C(C=C(C(=C1F)C=O)F)Cl