accessibility menu, dialog, popup

UserName

CH3NH3CH3CCH3OH

CAS RN 25400-83-3

IUPAC Name: 5-(dimethylamino)-2-methylpent-3-yn-2-ol
Synonyms: 5-(dimethylamino)-2-methylpent-3-yn-2-ol
Molecular Weight (g/mol): 141.21
Molecular Formula: C8H15NO
InChi Key: VBARYRPUABJYGW-UHFFFAOYSA-N
SMILES: CN(C)CC#CC(C)(C)O
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results