accessibility menu, dialog, popup

UserName

2H2H2H2H2H2H2H2H2H2H

CAS RN 25837-05-2

IUPAC Name: 1-[(1,1,2,2,2-²H₅)ethyl](²H₅)benzene
Synonyms: 1-[(1,1,2,2,2-²H₅)ethyl](²H₅)benzene
Molecular Weight (g/mol): 116.23
Molecular Formula: C8H10
InChi Key: YNQLUTRBYVCPMQ-CFTAVCBPSA-N
SMILES: [2H]C1=C([2H])C([2H])=C(C([2H])=C1[2H])C([2H])([2H])C([2H])([2H])[2H]
Molecular Weight (g/mol) 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)