accessibility menu, dialog, popup

UserName

CH3CH3OOOH3CCH3

CAS RN 26002-80-2

IUPAC Name: (3-phenoxyphenyl)methyl 2,2-dimethyl-3-(2-methylprop-1-en-1-yl)cyclopropane-1-carboxylate
Synonyms: phenothrin phenothrin (product) multicide 2154 epa pesticide chemical code 069005 (1R-trans)-phenothrin (1s-cis)-isomer phenothrin (1r-trans)-isomer phenothrin phenothrin (substance) phenothrin, (1r-cis)-isomer phenothrin, (1R-cis)-isomer
Molecular Weight (g/mol): 350.46
Molecular Formula: C23H26O3
InChi Key: SBNFWQZLDJGRLK-UHFFFAOYNA-N
SMILES: CC(C)=CC1C(C(=O)OCC2=CC=CC(OC3=CC=CC=C3)=C2)C1(C)C
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Grade 
  • (1)
Industry Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results