accessibility menu, dialog, popup

UserName

ClClOBr

CAS RN 2632-10-2

IUPAC Name: 2-bromo-1-(3,4-dichlorophenyl)ethan-1-one
Synonyms: 2-bromo-1-(3,4-dichlorophenyl)ethanone
Molecular Weight (g/mol): 267.93
Molecular Formula: C8H5BrCl2O
InChi Key: PAKFHEFMTRCFAU-UHFFFAOYSA-N
SMILES: ClC1=CC=C(C=C1Cl)C(=O)CBr
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results