CAS RN 26347-98-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
3
of
3
results
1
3,5-Bis(1,1-dimethylethyl)-4-hydroxy-3-hydroxy-2,2-bis(hydroxymethyl)propyl Benzenepropanoic Acid Ester, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
3,5-Bis(1,1-dimethylethyl)-4-hydroxy-3-hydroxy-2,2-bis(hydroxymethyl)propyl Benzenepropanoic Acid Ester, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
3,5-Bis(1,1-dimethylethyl)-4-hydroxy-3-hydroxy-2,2-bis(hydroxymethyl)propyl Benzenepropanoic Acid Ester, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.