accessibility menu, dialog, popup

UserName

H2N

CAS RN 2642-98-0

IUPAC Name: chrysen-6-amine
Synonyms: 6-aminochrysene
Molecular Weight (g/mol): 243.31
Molecular Formula: C18H13N
InChi Key: KIVUHCNVDWYUNP-UHFFFAOYSA-N
SMILES: NC1=CC2=C(C=CC3=CC=CC=C23)C2=CC=CC=C12
Quantity 
  • (1)
  • (1)
Color 
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results