accessibility menu, dialog, popup

UserName

FFOOHNH2

CAS RN 266360-63-8

IUPAC Name: 2-amino-3-(3,5-difluorophenyl)propanoic acid
Synonyms: 2-amino-3-(3,5-difluorophenyl)propanoic acid
Molecular Weight (g/mol): 201.17
Molecular Formula: C9H9F2NO2
InChi Key: QFGMPXZFCIHYIR-UHFFFAOYNA-N
SMILES: NC(CC1=CC(F)=CC(F)=C1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results