CAS RN 26788-23-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
4-Methoxy 17Beta-Estradiol, TRC
CAS: 26788-23-8 Molecular Formula: C19 H26 O3 Molecular Weight (g/mol): 302.41 Synonym: Estra-1,3,5(10)-triene-3,17-diol, 4-methoxy-, (17beta)-,Estra-1,3,5(10)-triene-3,17beta-diol, 4-methoxy- (8CI),(17beta)-4-Methoxyestra-1,3,5(10)-triene-3,17-diol,4-Methoxy-17beta-estradiol,4-Methoxyestradiol IUPAC Name: (8R,9S,13S,14S,17S)-4-methoxy-13-methyl-6,7,8,9,11,12,14,15,16,17-decahydrocyclopenta[a]phenanthrene-3,17-diol SMILES: COc1c(O)ccc2[C@H]3CC[C@]4(C)[C@@H](O)CC[C@H]4[C@@H]3CCc12