CAS RN 2680527-91-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
N-Nitroso Duloxetine, TRC
CAS: 2680527-91-5 Molecular Formula: C18 H18 N2 O2 S Molecular Weight (g/mol): 326.413 Synonym: 2-Thiophenepropanamine, N-methyl-γ-(1-naphthalenyloxy)-N-nitroso- (ACI),N-Methyl-N-[3-(1-naphthyloxy)-3-thien-2-ylpropyl]-N-nitrosoamine,N-Methyl-γ-(1-naphthalenyloxy)-N-nitroso-2-thiophenepropanamine,N-Nitroso-(±)-duloxetine,rac-N-Nitrosoduloxetine IUPAC Name: N-methyl-N-(3-naphthalen-1-yloxy-3-thiophen-2-ylpropyl)nitrous amide SMILES: CN(CCC(Oc1cccc2ccccc12)c3cccs3)N=O