accessibility menu, dialog, popup

UserName

CAS RN 302-01-2

NH2H2N

CAS RN 302-01-2

IUPAC Name: hydrazine
Synonyms: hydrazine diamine (specific compound) hydrazine base salts of hydrazine hydrazine, its derivatives and their salts anhydrous hydrazine hydrazine sulfate (product) nitrogen hydride diamine ccris 335
Molecular Weight (g/mol): 32.05
Molecular Formula: H4N2
InChi Key: OAKJQQAXSVQMHS-UHFFFAOYSA-N
SMILES: NN
Molecular Weight (g/mol) 
  • (1)
  • (4)
Solvent or Matrix 
  • (4)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results