CAS RN 314-40-9
CAS RN 314-40-9
IUPAC Name:
5-bromo-3-(butan-2-yl)-6-methyl-1,2,3,4-tetrahydropyrimidine-2,4-dione
Synonyms:
bromacil
Molecular Weight (g/mol):
261.12
Molecular Formula:
C9H13BrN2O2
InChi Key:
CTSLUCNDVMMDHG-UHFFFAOYNA-N
SMILES:
CCC(C)N1C(=O)NC(C)=C(Br)C1=O
1
–
6
of
6
results
3
Bromacil, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
4
Bromacil, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.