CAS RN 3857-25-8
CAS RN 3857-25-8
IUPAC Name:
(5-methylfuran-2-yl)methanol
Synonyms:
(5-methylfuran-2-yl)methanol
Molecular Weight (g/mol):
112.13
Molecular Formula:
C6H8O2
InChi Key:
VOZFDEJGHQWZHU-UHFFFAOYSA-N
SMILES:
CC1=CC=C(CO)O1
1
–
4
of
4
results
2
5-Methyl-2-furanmethanol, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
5-Methyl-2-furanmethanol, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
4
5-Methyl-2-furanmethanol, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.