accessibility menu, dialog, popup

UserName

OHClClNOO

CAS RN 39489-77-5

IUPAC Name: 2,4-dichloro-5-nitrophenol
Synonyms: 2,4-dichloro-5-nitrophenol
Molecular Weight (g/mol): 207.99
Molecular Formula: C6H3Cl2NO3
InChi Key: OFAPWTOOMSVMIU-UHFFFAOYSA-N
SMILES: OC1=CC(=C(Cl)C=C1Cl)[N+]([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results