accessibility menu, dialog, popup

UserName

OHOIF

CAS RN 403-18-9

IUPAC Name: 4-fluoro-3-iodobenzoic acid
Synonyms: 4-fluoro-3-iodobenzoic acid
Molecular Weight (g/mol): 266.01
Molecular Formula: C7H4FIO2
InChi Key: IKSOHJRYPQRTLQ-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC(I)=C(F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results