CAS RN 410536-97-9
IUPAC Name:
2-({[(1Z)-2-oxo-1,2-dihydronaphthalen-1-ylidene]methyl}amino)-N-[(1S)-1-phenylethyl]benzamide
Synonyms:
2-({[(1Z)-2-oxonaphthalen-1-ylidene]methyl}amino)-N-[(1S)-1-phenylethyl]benzamide
Molecular Weight (g/mol):
394.47
Molecular Formula:
C26H22N2O2
InChi Key:
YUGODMKHHCZZOI-ZVTCDHROSA-N
SMILES:
C[C@H](NC(=O)C1=CC=CC=C1N\C=C1/C(=O)C=CC2=CC=CC=C12)C1=CC=CC=C1
1
–
4
of
4
results
2
3
Sirtinol, MedChemExpress
MedChemExpress Sirtinol is a sirtuin (SIRT) inhibitor, with IC50s of 48 μM, 57.7 μM and 131 μM for ySir2, hSIRT2 and hSIRT2, respectively.
4
Sirtinol, R&D Systems™
CAS: 410536-97-9 Molecular Formula: C26H22N2O2 Molecular Weight (g/mol): 394.47 MDL Number: MFCD00810186 InChI Key: YUGODMKHHCZZOI-ZVTCDHROSA-N Synonym: 2-e-2-hydroxynaphthalen-1-yl methylidene amino-n-1-phenylethyl benzamide PubChem CID: 19577289 IUPAC Name: 2-({[(1Z)-2-oxo-1,2-dihydronaphthalen-1-ylidene]methyl}amino)-N-[(1S)-1-phenylethyl]benzamide SMILES: C[C@H](NC(=O)C1=CC=CC=C1N\C=C1/C(=O)C=CC2=CC=CC=C12)C1=CC=CC=C1