CAS RN 420-12-2
IUPAC Name:
thiirane
Synonyms:
thiirane aethylensulfid ethylene episulfide ethylene episulphide ccris 782
Molecular Weight (g/mol):
60.11
Molecular Formula:
C2H4S
InChi Key:
VOVUARRWDCVURC-UHFFFAOYSA-N
SMILES:
C1CS1
1
–
1
of
1
results
1
Thiirane, TRC
CAS: 420-12-2 Molecular Formula: C2H4S Molecular Weight (g/mol): 60.12 Synonym: Ethylene Sulfide,Ethylene episulfide,NSC 89690,Thiacyclopropane,2,3-Dihydro-thiirene IUPAC Name: thiirane SMILES: C1CS1