accessibility menu, dialog, popup

UserName

H3COCH3Br

CAS RN 42330-11-0

IUPAC Name: 4-bromononan-5-one
Synonyms: 4-bromononan-5-one
Molecular Weight (g/mol): 221.14
Molecular Formula: C9H17BrO
InChi Key: XNHZLGIYTMXPSA-UHFFFAOYNA-N
SMILES: CCCCC(=O)C(Br)CCC
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results