accessibility menu, dialog, popup

UserName

CAS RN 4330-99-8

CH3NH3CCH3NSCH3NH3CCH3NSOH(R)(R)OHHOOOHO

CAS RN 4330-99-8

IUPAC Name: (2R,3R)-2,3-dihydroxybutanedioic acid; bis(dimethyl[2-methyl-3-(10H-phenothiazin-10-yl)propyl]amine)
Synonyms: L(+)-tartaric acid; bis(trimeprazine)
Molecular Weight (g/mol): 746.98
Molecular Formula: C40H50N4O6S2
InChi Key: AJZJIYUOOJLBAU-CEAXSRTFSA-N
SMILES: O[C@H]([C@@H](O)C(O)=O)C(O)=O.CC(CN(C)C)CN1C2=CC=CC=C2SC2=CC=CC=C12.CC(CN(C)C)CN1C2=CC=CC=C2SC2=CC=CC=C12
Molecular Weight (g/mol) 
  • (3)
  • (3)
Certifications/Compliance 
  • (1)
  • (1)
  • (1)
Color 
  • (3)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (1)
  • (1)
  • (2)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)