CAS RN 4358-87-6
IUPAC Name:
methyl 2-hydroxy-2-phenylacetate
Synonyms:
methyl 2-hydroxy-2-phenylacetate
Molecular Weight (g/mol):
166.18
Molecular Formula:
C9H10O3
InChi Key:
ITATYELQCJRCCK-UHFFFAOYNA-N
SMILES:
COC(=O)C(O)C1=CC=CC=C1
1
–
2
of
2
results
2
Methyl (2RS)-2-Hydroxy-2-phenylacetate (Methyl Mandelate), Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.