accessibility menu, dialog, popup

UserName

CAS RN 475207-59-1

CH3HNOONHOHNClFFFNCH3SOHOO

CAS RN 475207-59-1

IUPAC Name: 4-[4-({[4-chloro-3-(trifluoromethyl)phenyl]carbamoyl}amino)phenoxy]-N-methylpyridine-2-carboxamide; 4-methylbenzene-1-sulfonic acid
Synonyms: para-toluene sulfonate; sorafenib
Molecular Weight (g/mol): 637.03
Molecular Formula: C28H24ClF3N4O6S
InChi Key: IVDHYUQIDRJSTI-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)S(O)(=O)=O.CNC(=O)C1=CC(OC2=CC=C(NC(=O)NC3=CC=C(Cl)C(=C3)C(F)(F)F)C=C2)=CC=N1
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Certifications/Compliance 
  • (1)
  • (1)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)