CAS RN 51-98-9
CAS RN 51-98-9
IUPAC Name:
(1R,3aS,3bR,9aR,9bS,11aS)-1-ethynyl-11a-methyl-7-oxo-1H,2H,3H,3aH,3bH,4H,5H,7H,8H,9H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-1-yl acetate
Synonyms:
ENTA norlutin A miniphase progylut norethysterone acetate milli-anovlar monogest gestakadin norethisteron acetate primolut-nor
Molecular Weight (g/mol):
340.46
Molecular Formula:
C22H28O3
InChi Key:
IMONTRJLAWHYGT-ZCPXKWAGSA-N
SMILES:
CC(=O)O[C@]1(CC[C@H]2[C@@H]3CCC4=CC(=O)CC[C@@H]4[C@H]3CC[C@]12C)C#C
1
–
3
of
3
results
1
Norethisterone Acetate, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2
Norethisterone Acetate (Norethindrone Acetate), LoGiCal
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
3
Norethindrone acetate, MedChemExpress
MedChemExpress Norethindrone acetate is a female hormone used for the research of endometriosis.