accessibility menu, dialog, popup

UserName

CH3NSNSH3CI(E/Z)(E/Z)

CAS RN 53336-12-2

IUPAC Name: 3-propyl-2-[3-(3-propyl-2,3-dihydro-1,3-benzothiazol-2-ylidene)prop-1-en-1-yl]-1,3-benzothiazol-3-ium iodide
Synonyms: 3-propyl-2-[3-(3-propyl-1,3-benzothiazol-2-ylidene)prop-1-en-1-yl]-1,3-benzothiazol-3-ium iodide
Molecular Weight (g/mol): 520.49
Molecular Formula: C23H25IN2S2
InChi Key: JGTCEHAVVINOPG-UHFFFAOYSA-M
SMILES: [I-].CCCN1C(SC2=CC=CC=C12)=CC=CC1=[N+](CCC)C2=CC=CC=C2S1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results