Learn More
3,3'-Di-n-propylthiacarbocyanine iodide, 96%, Thermo Scientific Chemicals
Product Code. 15415127
Change view
Click to view available options
Quantity:
250 mg
5 g
Unit Size:
250mg
5g
Product Code. Quantity unitSize
15415127 5 g 5g
15425127 250 mg 250mg
2 options
Product Code. 15415127
Brand: Thermo Scientific Alfa Aesar H32400.06
Request Bulk or Custom Format
This item requires a price update prior to being ordered.
Request a Price for this item here.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 53336-12-2
Molecular Formula C23H25IN2S2
Molecular Weight (g/mol) 520.49
MDL Number MFCD00013340
InChI Key JGTCEHAVVINOPG-UHFFFAOYSA-M
Synonym 3,3'-dipropylthiacarbocyanine iodide, 3-propyl-2-3-3-propyl-2 3h benzothiazolylidene-1-propenyl benzothiazolium iodide, 3-propyl-2-1e,3z-3-3-propylbenzo d thiazol-2 3h-ylidene prop-1-en-1-yl benzo d thiazol-3-ium iodide
PubChem CID 57369736
SMILES [I-].CCCN1C(SC2=CC=CC=C12)=CC=CC1=[N+](CCC)C2=CC=CC=C2S1
Specifications
Quantity 5 g
IUPAC Name 3-propyl-2-[3-(3-propyl-1,3-benzothiazol-3-ium-2-yl)prop-2-enylidene]-1,3-benzothiazole;iodide
Formula Weight 520.5
Percent Purity 96%
Chemical Name or Material 3,3'-Di-n-propylthiacarbocyanine iodide