CAS RN 5538-41-0
CAS RN 5538-41-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
3
of
3
results
1
2-Chloro-3,5-dimethylphenol, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2
2-Chloro-3,5-dimethylphenol, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
2-Chloro-3,5-dimethylphenol, TRC
CAS: 5538-41-0 Molecular Formula: C8 H9 Cl O Molecular Weight (g/mol): 156.61 Synonym: Phenol, 2-chloro-3,5-dimethyl-,3,5-Xylenol, 2-chloro- (7CI,8CI),2-Chloro-3,5-dimethylphenol,2-Chloro-3,5-xylenol IUPAC Name: 2-chloro-3,5-dimethylphenol SMILES: Cc1cc(C)c(Cl)c(O)c1