accessibility menu, dialog, popup

UserName

CAS RN 6011-14-9

ClNH3N

CAS RN 6011-14-9

IUPAC Name: cyanomethanaminium chloride
Synonyms: cyanomethanaminium chloride
Molecular Weight (g/mol): 92.53
Molecular Formula: C2H5ClN2
InChi Key: XFKYKTBPRBZDFG-UHFFFAOYSA-N
SMILES: [Cl-].[NH3+]CC#N
Molecular Weight (g/mol) 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results