accessibility menu, dialog, popup

UserName

CAS RN 6211-32-1

ClCH3OO(S)HO(S)NHN(S)(S)H

CAS RN 6211-32-1

IUPAC Name: hydrogen methyl (1S,15S,18S,20S)-18-hydroxy-3,13-diazapentacyclo[11.8.0.0²,¹⁰.0⁴,⁹.0¹⁵,²⁰]henicosa-2(10),4,6,8-tetraene-19-carboxylate chloride
Synonyms: hydrogen methyl (1S,15S,18S,20S)-18-hydroxy-3,13-diazapentacyclo[11.8.0.0²,¹⁰.0⁴,⁹.0¹⁵,²⁰]henicosa-2(10),4,6,8-tetraene-19-carboxylate chloride
Molecular Weight (g/mol): 390.91
Molecular Formula: C21H27ClN2O3
InChi Key: PIPZGJSEDRMUAW-MIFCKTTJSA-N
SMILES: [H+].[Cl-].COC(=O)C1[C@@H](O)CC[C@@H]2CN3CCC4=C(NC5=CC=CC=C45)[C@@H]3C[C@H]12
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)