accessibility menu, dialog, popup

UserName

CH3NH2HOPOHOOHNBrCl

CAS RN 6578-06-9

IUPAC Name: 4-methylaniline; [(5-bromo-4-chloro-1H-indol-3-yl)oxy]phosphonic acid
Synonyms: (5-bromo-4-chloro-1H-indol-3-yl)oxyphosphonic acid; toluidine
Molecular Weight (g/mol): 433.62
Molecular Formula: C15H15BrClN2O4P
InChi Key: QEIFSLUFHRCVQL-UHFFFAOYSA-N
SMILES: CC1=CC=C(N)C=C1.OP(O)(=O)OC1=CNC2=CC=C(Br)C(Cl)=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results