CAS RN 65872-41-5
CAS RN 65872-41-5
IUPAC Name:
(2Z)-2-(2-amino-1,3-thiazol-4-yl)-2-(methoxyimino)acetate
Synonyms:
(2Z)-2-(2-amino-1,3-thiazol-4-yl)-2-(methoxyimino)acetate
Molecular Weight (g/mol):
200.19
Molecular Formula:
C6H6N3O3S
InChi Key:
NLARCUDOUOQRPB-WTKPLQERSA-M
SMILES:
CO\N=C(/C([O-])=O)C1=CSC(N)=N1
1
–
4
of
4
results
1
2-Amino-a-(methoxyimino)-4-thiazoleacetic Acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
(2Z)-(2-Aminothiazol-4-yl)(methoxyimino)acetic Acid, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
3
2-Amino-a-(methoxyimino)-4-thiazoleacetic Acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
4
2-Amino-Alpha-(methoxyimino)-4-thiazoleacetic Acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.