CAS RN 67905-19-5
CAS RN 67905-19-5
IUPAC Name:
hentriacontafluorohexadecanoic acid
Synonyms:
hentriacontafluorohexadecanoic acid
Molecular Weight (g/mol):
814.13
Molecular Formula:
C16HF31O2
InChi Key:
OJMBMWRMTMHMSZ-UHFFFAOYSA-N
SMILES:
OC(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F
1
–
1
of
1
results
1
Perfluorohexadecanoic acid 50 μg/mL in Methanol:Water, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality