CAS RN 7280-81-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
3-Benzyladenine, TRC
CAS: 7280-81-1 Molecular Formula: C12H11N5 Molecular Weight (g/mol): 225.25 IUPAC Name: 3-benzyl-7H-purin-6-imine SMILES: Nc1ncn(Cc2ccccc2)c3ncnc13